Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPHA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588016 Supplier Invitrogen™ Supplier No. PA588016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QEAVIPGCHL HPFNVTVRSD RQGTCQGSHV AQACVGHCES SAFPSRYSVL VASGYRHNIT SVSQCCTISG LKKVKVQLQC VGSRREELEI FTARACQCDM CRLSRY; Positive Samples: Rat brain.

GPHA2 is a cystine knot-forming polypeptide and a subunit of the dimeric glycoprotein hormone family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPHA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Gpha2
Gene Accession No. Q925Q4
Gene Alias A2; cysteine knot protein; glycoprotein alpha 2; glycoprotein hormone alpha 2; glycoprotein hormone alpha-2; Gpa2; Gpha2; Putative secreted protein Zsig51; thyrostimulin subunit alpha; ZSIG51
Gene Symbols Gpha2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 24-129 of human GPHA2 (NP_5701251).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171158
Target Species Rat, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.