Learn More
Invitrogen™ GPHB5 Polyclonal Antibody

Description
GPHB5 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | GPHB5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | GPHB5 |
| Gene Accession No. | Q86YW7 |
| Gene Alias | B5; glycoprotein beta 5; glycoprotein hormone beta 5; glycoprotein hormone beta subunit; glycoprotein hormone beta-5; GPB5; GPHB5; Thyrostimulin subunit beta; ZLUT1 |
| Gene Symbols | GPHB5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.