Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPI Recombinant Rabbit Monoclonal Antibody (2Y8R0)
GREENER_CHOICE

Catalog No. PIMA535506
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535506 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535506 Supplier Invitrogen™ Supplier No. MA535506
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MAALTRDPQF QKLQQWYREH RSELNLRRLF DANKDRFNHF SLTLNTNHGH ILVDYSKNLV TEDVMRMLVD LAKSRGVEAA RERMFNGEKI NYTEGRAVLH.

This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPI
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2Y8R0
Concentration 0.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPI
Gene Accession No. P06744
Gene Alias Amf; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250; GNPI; Gpi; GPI {ECO:0000250; gpi {ECO:0000312; GPI1; Gpi-1; Gpi-1r; Gpi1-r; Gpi1s; Gpi-1s; Gpi1-s; Gpi-1t; Gpi1-t; GPI-GlcNAc transferase complex subunit GPI1; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; MF; Neuroleukin; neuroleukin {ECO:0000250; NK; NK/GPI; Nlk; NLK {ECO:0000250; Org; oxoisomerase; PGI; PGI {ECO:0000250; Phi; PHI {ECO:0000250; phosphatidylinositol N-acetylglucosaminyltransferase; Phosphatidylinositol N-acetylglucosaminyltransferase subunit GPI1; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250; phosphohexomutase; phosphohexose isomerase; phosphohexose isomerase {ECO:0000250; phosphosaccharomutase; RGD:2727}; SA36; SA-36; sperm antigen; sperm antigen 36; sperm antigen-36; UniProtKB:P06745}; YGR216C
Gene Symbols GPI
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Glucose 6 phosphate isomerase (P06744).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2821
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.