Learn More
GPI Mouse anti-Human, Clone: OTI7G10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501136

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.Specifications
GPI | |
Monoclonal | |
Unconjugated | |
GPI | |
AMF, GNPI, NLK, PGI, PHI, SA-36, SA36 | |
Mouse | |
Affinity Chromatography | |
RUO | |
2821 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7G10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06744 | |
GPI | |
Full length human recombit protein of human GPI produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.