Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPI Mouse anti-Human, Unconjugated, Clone: CPTC-GPI-1, Abnova™

Catalog No. 89337516 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-516 Supplier Abnova Corporation Supplier No. MAB21023
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GPI Monoclonal antibody specifically detects GPI in Human samples. It is validated for WB-Ce, IHC-P, IF.

Specifications

Antigen GPI
Applications Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Monoclonal
Clone CPTC-GPI-1
Conjugate Unconjugated
Description glucose phosphate isomerase
Gene GPI
Gene Alias AMF, GNPI, NLK, PGI, PHI, SA-36
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human GPI
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2821
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.