Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597517
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597517 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597517 Supplier Invitrogen™ Supplier No. PA597517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPI Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPI
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GPI
Gene Accession No. P06744, P06745
Gene Alias Amf; Autocrine motility factor; glucose phosphate isomerase; glucose phosphate isomerase 1; glucose phosphate isomerase 1 complex; glucose phosphate isomerase 1, regulatory; glucose phosphate isomerase 1, structural; glucose phosphate isomerase 1, temporal; glucose-6-phosphate isomerase; glucose-6-phosphate isomerase {ECO:0000250; GNPI; Gpi; GPI {ECO:0000250; gpi {ECO:0000312; GPI1; Gpi-1; Gpi-1r; Gpi1-r; Gpi1s; Gpi-1s; Gpi1-s; Gpi-1t; Gpi1-t; GPI-GlcNAc transferase complex subunit GPI1; hexose monophosphate isomerase; hexosephosphate isomerase; maturation factor; MF; Neuroleukin; neuroleukin {ECO:0000250; NK; NK/GPI; Nlk; NLK {ECO:0000250; Org; oxoisomerase; PGI; PGI {ECO:0000250; Phi; PHI {ECO:0000250; phosphatidylinositol N-acetylglucosaminyltransferase; Phosphatidylinositol N-acetylglucosaminyltransferase subunit GPI1; Phosphoglucose isomerase; phosphoglucose isomerase {ECO:0000250; phosphohexomutase; phosphohexose isomerase; phosphohexose isomerase {ECO:0000250; phosphosaccharomutase; RGD:2727}; SA36; SA-36; sperm antigen; sperm antigen 36; sperm antigen-36; UniProtKB:P06745}; YGR216C
Gene Symbols GPI, Gpi1
Host Species Rabbit
Immunogen Recombinant Human Glucose-6-phosphate isomerase protein (2-554AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14751, 2821
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.