Learn More
GPI Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | GPI |
---|---|
Applications | Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-5966
![]() |
Bethyl Laboratories, Inc
A303753AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This gene belongs to the GPI family whose members encode multifunctional phosphoglucose isomerase proteins involved in energy pathways. The protein encoded by this gene is a dimeric enzyme that catalyzes the reversible isomerization of glucose-6-phosphate and fructose-6-phosphate. The protein functions in different capacities inside and outside the cell. In the cytoplasm, the gene product is involved in glycolysis and gluconeogenesis, while outside the cell it functions as a neurotrophic factor for spinal and sensory neurons. Defects in this gene are the cause of nonspherocytic hemolytic anemia and a severe enzyme deficiency can be associated with hydrops fetalis, immediate neonatal death and neurological impairment.Specifications
GPI | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
GPI; glucose phosphate isomerase; AMF; GNPI; NLK; PGI; PHI; SA-36; autocrine motility factor; glucose-6-phosphate isomerase; hexose monophosphate isomerase; hexosephosphate isomerase; neuroleukin; oxoisomerase; phosphoglucose isomerase; phosphohexomutase; phosphohexose isomerase; phosphosaccharomutase; sperm antigen-36 | |
GPI | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P06744 | |
2821 | |
Between 508 and 558 | |
Primary | |
4° C | |
GPI |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.