Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPI8 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299379V
Description
GPI8 Polyclonal specifically detects GPI8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GPI8 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
10026 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 405 | |
EC 3.-, GPI transamidase, GPI transamidase subunit, GPI8 homolog, GPI8class K, GPI-anchor transamidase, hGPI8, MGC22559, phosphatidylinositol glycan anchor biosynthesis, class K, Phosphatidylinositol-glycan biosynthesis class K protein, PIG-K | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human GPI8. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction