Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPIHBP1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297041AF350
Description
GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for ELISA.Specifications
GPIHBP1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
Alexa Fluor 350 | |
glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
Produced in rabbits immunized with purified, recombinant GPIHBP1 (AAH63857.1; Met1-Ser160). | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
338328 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction