Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPIHBP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11041539AF488
Description
GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
GPIHBP1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GPIHBP1 | |
A synthetic peptide corresponding to an internal region [within residues 170-250] of mouse GPIHBP1. [Swiss-Prot# Q9D1N2] | |
0.1 mL | |
Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction | |
338328 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction