Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPIHBP1 Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11055293B
Description
GPIHBP1 Polyclonal specifically detects GPIHBP1 in Human samples. It is validated for Western Blot.Specifications
| GPIHBP1 | |
| Polyclonal | |
| Western Blot | |
| Q8IV16 | |
| GPIHBP1 | |
| A synthetic peptide corresponding to an internal region [within residues 100-180] of human GPIHBP1. [Swiss-Prot# Q8IV16] | |
| 0.1 mL | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 338328 | |
| Human | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Immunogen has 78% homology to mouse. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction