Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPIHBP1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$207.00 - $499.50

Specifications

Antigen GPIHBP1
Concentration 0.9 mg/mL
Dilution Western Blot 2.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:50
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11041539T
SDP
View Documents
Novus Biologicals
NB11041539SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB11041539
SDP
View Documents
Novus Biologicals
NB11041539
0.1 mL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

GPIHBP1 Polyclonal antibody specifically detects GPIHBP1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GPIHBP1
Western Blot 2.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:20 - 1:50
Polyclonal
Rabbit
Lipid and Metabolism, Signal Transduction
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
338328
A synthetic peptide corresponding to an internal region [within residues 170-250] of mouse GPIHBP1. [Swiss-Prot# Q9D1N2]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
0.9 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Mouse
glycosylphosphatidylinositol anchored high density lipoprotein binding protein1, glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein1, GPI anchored high density lipoprotein binding protein 1, GPI-anchored HDL-binding protein 1, GPI-HBP1LOC338328, HBP1, High density lipoprotein-binding protein 1
GPIHBP1
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.