Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPIHBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545576
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545576 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545576 Supplier Invitrogen™ Supplier No. PA545576
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QVTMTCCQSS LCNVPPWQSS RVQDPTGKGA GGPRGSSETV GAALLLNLLA Sequence homology: Human: 100%.

GPI-anchored HDL-binding protein 1 (Gpihbp1) is a novel protein that may be involved in the regulation of the delivery of fats to cells for energy and storage. Digested fats travel to the small intestine, where they are packaged into chylomicrons (particles filled with triglycerides). Chylomicrons then travel through the bloodstream and deliver triglycerides to tissues that are hungry for fuel or to adipose tissue for energy storage. Triglycerides are broken down or hydrolyzed by the enzyme lipoprotein lipase (LpL). The triglyceride breakdown products are then taken up and used by cells. Gpihbp1 is the molecule in capillaries that facilitates the capture of chylomicrons and facilitates the interaction with LpL. It has been shown that fats in the bloodstream are not readily metabolized in the absence of GPIHBP1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPIHBP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GPIHBP1
Gene Accession No. Q8IV16
Gene Alias 1110002J19Rik; endothelial cell LPL transporter; glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1; glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI anchored high density lipoprotein binding protein 1; GPI-anchored HDL-binding protein 1; Gpihbp1; GPI-HBP1; Hbp1; HDLBP; HDLBP1; High density lipoprotein-binding protein 1; HYPL1D; RGD1564237
Gene Symbols GPIHBP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GPIHBP1 (aa 125-174).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338328
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.