Learn More
Invitrogen™ NSG2 Polyclonal Antibody

Description
Immunogen sequence: RCIPASLDAY YSSQDPNSRS RFYTVISHYS VAKQSTARAI GPWLSAAAVI HEPKPPKTQG H Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Specifications
Specifications
| Antigen | NSG2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NSG2 |
| Gene Accession No. | Q9Y328 |
| Gene Alias | CALY3; HMP19; HMP19 protein; hypothalamus golgi apparatus expressed 19 kDa protein; neuron specific gene family member 2; Neuronal vesicle trafficking-associated protein 2; neuron-specific protein family member 2; NSG2; p19 protein; protein 8.5; Protein p19 |
| Gene Symbols | NSG2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.