Learn More
Invitrogen™ GPIHBP1 Polyclonal Antibody

Description
Suggested positive control: mouse kidney lysate or protein.
Specifications
Specifications
| Antigen | GPIHBP1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.9 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% sodium azide |
| Gene | GPIHBP1 |
| Gene Accession No. | Q8IV16, Q9D1N2 |
| Gene Alias | 1110002J19Rik; endothelial cell LPL transporter; glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1; glycosylphosphatidylinositol-anchored high density lipoprotein-binding protein 1; GPI anchored high density lipoprotein binding protein 1; GPI-anchored HDL-binding protein 1; Gpihbp1; GPI-HBP1; Hbp1; HDLBP; HDLBP1; High density lipoprotein-binding protein 1; HYPL1D; RGD1564237 |
| Gene Symbols | GPIHBP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.