Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPIP137 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222238
Description
GPIP137 Polyclonal specifically detects GPIP137 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
GPIP137 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
activation/proliferation-associated protein 1, caprin 1, caprin-1, cell cycle associated protein 1, Cell cycle-associated protein 1, Cytoplasmic activation- and proliferation-associated protein 1, cytoplasmic activation/proliferation-associated protein-1, GPI-anchored membrane protein 1GPIP137, GPI-p137, M11S1GPI-anchored protein p137, Membrane component chromosome 11 surface marker 1, membrane component, chromosome 11, surface marker 1, p137GPI, RNA granule protein 105, RNG105GPIAP1 | |
Rabbit | |
Affinity Purified | |
RUO | |
4076 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
NP_005889.3 | |
CAPRIN1 | |
The immunogen this antibody was made to, maps to a region between residue 550 and 600 of human Cell Cycle Associated Protein 1 using the numbering given in entry NP_005889.3 (GeneID 4076). | |
100 μL | |
Primary | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction