Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPIP137 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA5143227
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_005889.3; NP_976240.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
GPIP137 regulates the transport and translation of mRNAs of proteins involved in synaptic plasticity in neurons and cell proliferation/migration in multiple cell types. It binds directly and selectively to MYC and CCND2 RNAs. In neuronal cells, GPIP137 binds to several mRNAs associated with RNA granules including BDNF, CAMK2A, CREB1, MAP2, NTRK2, mRNAs, GRIN1, and KPNB1 mRNAs.
Specifications
| GPIP137 | |
| Polyclonal | |
| Unconjugated | |
| Caprin1 | |
| activation/proliferation-associated protein 1; AL022980; caprin 1; Caprin1; caprin-1; cell cycle associated protein 1; cell cycle-associated protein 1; cytop; Cytoplasmic activation- and proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein 1; cytoplasmic activation/proliferation-associated protein-1; G5E5; GPI p137; GPI-anchored membrane protein 1; GPI-anchored protein p137; Gpiap; GPIAP1; Gpip137; GPI-p137; GRIP137; M11S1; Membrane component chromosome 11 surface marker 1; membrane component, chromosome 11, surface marker 1; Mmgpip137; P137; p137GPI; RNA granule protein 105; Rng105 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4076 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q14444 | |
| Caprin1 | |
| Peptide with sequence C-EQRPQKEPIDQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction