Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPKOW Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$405.00 - $670.00
Specifications
Antigen | GPKOW |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GPKOW Polyclonal specifically detects GPKOW in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GPKOW | |
Polyclonal | |
Rabbit | |
Human, Mouse, Rat | |
G patch domain and KOW motifs, GPATC5G patch domain and KOW motifs-containing protein, GPATCH5G patch domain-containing protein 5, Protein MOS2 homolog, Spp2, T54Protein T54 | |
GPKOW | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
27238 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:ELQSVKPQEAPKELVIPLIQNGHRRQPPARPPGPSTDTGALADGVVSQAVKELIAESKKSLEERENAGVDPTLAIPMIQKGCTPSGEGADSEPRAETVPEEANYEA | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title