Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPKOW Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531001
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
Specifications
| GPKOW | |
| Polyclonal | |
| Unconjugated | |
| GPKOW | |
| 4432411B18; DXErtd697e; DXImx41e; G patch domain and KOW motifs; G patch domain and KOW motifs-containing protein; GPATC5; G-patch domain and KOW motifs; G-patch domain and KOW motifs-containing protein; G-patch domain-containing protein 5; GPATCH5; GPKOW; Protein MOS2 homolog; Protein T54; Spp2; T54 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27238 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q92917 | |
| GPKOW | |
| Recombinant protein encompassing a sequence within the center region of human GPKOW. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction