Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPKOW Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551374
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551374 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551374 Supplier Invitrogen™ Supplier No. PA551374
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELQSVKPQEA PKELVIPLIQ NGHRRQPPAR PPGPSTDTGA LADGVVSQAV KELIAESKKS LEERENAGVD PTLAIPMIQK GCTPSGEGAD SEPRAETVPE EANYEA Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

This gene encodes a putative RNA-binding protein containing G-patch and KOW (Kyprides, Ouzounis, Woese) domains. The encoded protein interacts directly with protein kinase A and protein kinase X and is also found associated with the spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPKOW
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPKOW
Gene Accession No. Q56A08, Q92917
Gene Alias 4432411B18; DXErtd697e; DXImx41e; G patch domain and KOW motifs; G patch domain and KOW motifs-containing protein; GPATC5; G-patch domain and KOW motifs; G-patch domain and KOW motifs-containing protein; G-patch domain-containing protein 5; GPATCH5; GPKOW; Protein MOS2 homolog; Protein T54; Spp2; T54
Gene Symbols GPKOW
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPKOW. Recombinant protein control fragment (Product #RP-91888).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209416, 27238, 679890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.