Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104013 Supplier Invitrogen™ Supplier No. PA5104013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPLD1.

GPLD1 is expressed in numerous tissues and cells and specifically cleaves GPI-anchored proteins. Liver has the highest level of GPI-PLD expression and is the primary organ contributing to GPLD1 in the serum. GPLD1 is abundant in serum in which it associates with polipoproteins AI and AIV. Increased serum GPLD1 is associated with insulin resistance and elevated serum triglycerides. Many surface proteins are attached to eukaryotic cell membranes via glycosylphosphatidylinositol (GPI) anchors that are covalently bound to the C-terminus of the protein and cleavage of the GPI moiety by GPLD1, only enzyme known that cleavage GPI anchor, may represent a means of regulating attachment of these proteins to the cell surface, or alternatively, their release into the extracellular environment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPLD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPLD1
Gene Accession No. O70362, P80108
Gene Alias 6330541J12Rik; AW546131; glycoprotein phospholipase D; glycosylphosphatidylinositol phospholipase D; glycosylphosphatidylinositol specific phospholipase D1; glycosylphosphatidylinositol specific phospholipase D1, isoform 2; Glycosyl-phosphatidylinositol-specific phospholipase D; GPIPLD; GPI-PLD; GPIPLDM; GPI-specific phospholipase D; Gpld1; MGC22590; Phosphatidylinositol-glycan-specific phospholipase D; PI-G PLD; PIGPLD; PIGPLD1; PLD
Gene Symbols GPLD1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPLD1(Accession P80108), corresponding to amino acid residues F247-S297.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14756, 2822
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.