Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPM6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553588 Supplier Invitrogen™ Supplier No. PA553588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YFNLWTICRN TTLVEGANLC LDLRQFGIVT IGEEKKICTV SENFLRMCES TELN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 98%.

Neuronal membrane glycoprotein M6-a is a protein that in humans is encoded by the GPM6A gene. Involved in neuronal differentiation, including differentiation and migration of neuronal stem cells. Plays a role in neuronal plasticity and is involved in neurite and filopodia outgrowth, filopodia motility and probably synapse formation. GPM6A-induced filopodia formation involves mitogen-activated protein kinase (MAPK) and Src signaling pathways. May be involved in neuronal NGF-dependent Ca2+ influx. May be involved in regulation of endocytosis and intracellular trafficking of G-protein-coupled receptors (GPCRs); enhances internalization and recycling of mu-type opioid receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPM6A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPM6A
Gene Accession No. P51674
Gene Alias glycoprotein m6a; GPM6; Gpm6a; M6A; neuronal membrane glycoprotein M6-a; neuronal membrane glycoprotein M6-a-like
Gene Symbols GPM6A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPM6A. Recombinant protein control fragment (Product #RP-91159).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2823
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.