Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPM6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103972 Supplier Invitrogen™ Supplier No. PA5103972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPM6B.

May be involved in neural development. Involved in regulation of osteoblast function and bone formation. Involved in matrix vesicle release by osteoblasts; this function seems to involve maintenance of the actin cytoskeleton. May be involved in cellular trafficking of SERT and thereby in regulation of serotonin uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPM6B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPM6B
Gene Accession No. P35803, Q13491, Q9JJK1
Gene Alias AI593561; glycoprotein M6B; Gpm6; GPM6B; M6b; neuronal membrane glycoprotein M6-b; protolipid M6B; rhombencephalic expression protein-29 kDa; Rhombex-29
Gene Symbols GPM6B
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPM6B(Accession Q13491), corresponding to amino acid residues M1-S37.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14758, 192179, 2824
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.