Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPM6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112897
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112897 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112897 Supplier Invitrogen™ Supplier No. PA5112897
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPM6B Polyclonal Antibody for ICC/IF, IHC (P), ELISA

May be involved in neural development. Involved in regulation of osteoblast function and bone formation. Involved in matrix vesicle release by osteoblasts; this function seems to involve maintenance of the actin cytoskeleton. May be involved in cellular trafficking of SERT and thereby in regulation of serotonin uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPM6B
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 2.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene GPM6B
Gene Accession No. Q13491
Gene Alias AI593561; glycoprotein M6B; Gpm6; GPM6B; M6b; neuronal membrane glycoprotein M6-b; protolipid M6B; rhombencephalic expression protein-29 kDa; Rhombex-29
Gene Symbols GPM6B
Host Species Rabbit
Immunogen Recombinant Human Neuronal membrane glycoprotein M6-b protein (1-64AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2824
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.