Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPM6B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551773 Supplier Invitrogen™ Supplier No. PA551773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AVPVFMFYNI WSTCEVIKSP QTNGTTGVEQ ICVDIRQYGI IPWNAFPGKI CGSALENICN TNEFYMSYH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

May be involved in neural development. Involved in regulation of osteoblast function and bone formation. Involved in matrix vesicle release by osteoblasts; this function seems to involve maintenance of the actin cytoskeleton. May be involved in cellular trafficking of SERT and thereby in regulation of serotonin uptake.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPM6B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GPM6B
Gene Accession No. Q13491
Gene Alias AI593561; glycoprotein M6B; Gpm6; GPM6B; M6b; neuronal membrane glycoprotein M6-b; protolipid M6B; rhombencephalic expression protein-29 kDa; Rhombex-29
Gene Symbols GPM6B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPM6B. Recombinant protein control fragment (Product #RP-92163).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2824
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.