Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPM6B Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11847R
Description
May be involved in neural development. Involved in regulation of osteoblast function and bone formation. Involved in matrix vesicle release by osteoblasts; this function seems to involve maintenance of the actin cytoskeleton. May be involved in cellular trafficking of SERT and thereby in regulation of serotonin uptake.Specifications
| GPM6B | |
| Polyclonal | |
| Unconjugated | |
| GPM6B | |
| AI593561; glycoprotein m6b; Gpm6; Gpm6b; M6b; Neuronal membrane glycoprotein M6-b; protolipid M6B; Rhombencephalic expression protein-29 kDa; rhombex-29 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2824 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 μg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
| Q13491 | |
| GPM6B | |
| KLH conjugated synthetic peptide derived from human GPM6B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction