Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPN1 Mouse anti-Human, Unconjugated, Clone: GPN1/2350, Abnova™
Mouse Monoclonal Antibody.
Supplier: Abnova Corporation MAB20883
Description
GPN1 Monoclonal antibody specifically detects GPN1 in Human samples. It is validated for IHC-P.Specifications
GPN1 | |
Monoclonal | |
Unconjugated | |
GPN1 | |
Mouse | |
100 μg | |
11321 | |
Store at 2 to 8°C | |
IgG1 κ |
Immunohistochemistry (Paraffin) | |
GPN1/2350 | |
GPN-loop GTPase 1 | |
ATPBD1A, MBDIN, NTPBP, XAB1 | |
Recombinant protein corresponding to full length human GPN1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction