Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112073 Supplier Invitrogen™ Supplier No. PA5112073
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPN1 Polyclonal Antibody for Western Blot, IP

Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII) (PubMed:20855544, PubMed:21768307). May act at an RNAP assembly step prior to nuclear import (PubMed:21768307). Forms an interface between the RNA polymerase II enzyme and chaperone/scaffolding proteins, suggesting that it is required to connect RNA polymerase II to regulators of protein complex formation (PubMed:17643375). May be involved in nuclear localization of XPA (PubMed:11058119).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPN1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Gpn1
Gene Accession No. Q9HCN4
Gene Alias 2410004J02Rik; AI449615; ATP(GTP)-binding protein; ATPBD1A; GPN1; GPN-loop GTPase 1; HUSSY-23; MBD2-interacting protein; MBDIN; methyl-CpG binding domain protein 2 interacting protein; NTPBP; RNA polymerase II associated protein 4; RNAPII-associated protein 4; RPAP4; XAB1; XPA binding protein 1; XPA binding protein 1, GTPase; XPA-binding protein 1
Gene Symbols Gpn1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the C-terminus of the Human GPN1/XAB1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11321
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.