Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112202
Description
GPN1 Polyclonal Antibody for Western Blot, IP
Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII) (PubMed:20855544, PubMed:21768307). May act at an RNAP assembly step prior to nuclear import (PubMed:21768307). Forms an interface between the RNA polymerase II enzyme and chaperone/scaffolding proteins, suggesting that it is required to connect RNA polymerase II to regulators of protein complex formation (PubMed:17643375). May be involved in nuclear localization of XPA (PubMed:11058119).
Specifications
GPN1 | |
Polyclonal | |
Unconjugated | |
Gpn1 | |
2410004J02Rik; AI449615; ATP(GTP)-binding protein; ATPBD1A; GPN1; GPN-loop GTPase 1; HUSSY-23; MBD2-interacting protein; MBDIN; methyl-CpG binding domain protein 2 interacting protein; NTPBP; RNA polymerase II associated protein 4; RNAPII-associated protein 4; RPAP4; XAB1; XPA binding protein 1; XPA binding protein 1, GTPase; XPA-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
11321 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9HCN4 | |
Gpn1 | |
E. coli-derived Human XAB1/GPN1 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction