Learn More
GPN1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304615AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Small GTPase required for proper nuclear import of RNA polymerase II (RNAPII) (PubMed:20855544, PubMed:21768307). May act at an RNAP assembly step prior to nuclear import (PubMed:21768307). Forms an interface between the RNA polymerase II enzyme and chaperone/scaffolding proteins, suggesting that it is required to connect RNA polymerase II to regulators of protein complex formation (PubMed:17643375). May be involved in nuclear localization of XPA (PubMed:11058119).Specifications
GPN1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9HCN4 | |
GPN1 | |
Between 324 and 374 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GPN1 | |
GPN-loop GTPase 1, MBDIN, ATPBD1A, NTPBP, GPN1, RPAP4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11321 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.