Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPNMB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589716 Supplier Invitrogen™ Supplier No. PA589716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, K-562, U-87MG, Mouse brain, Mouse liver, Rat brain, Rat liver; Cellular Location: Cell membrane, Melanosome, Single-pass type I membrane protein Immunogen sequence: KRFHDVLGNE RPSAYMREHN QLNGWSSDEN DWNEKLYPVW KRGDMRWKNS WKGGRVQAVL TSDSPALVGS NITFAVNLIF PRCQKEDANG NIVYEKNCRN EAGLSADPYV YNWTAWSEDS DGENGTGQSH HNVFPDGKPF PHHPGWRRWN FIYVFHTLGQ YFQKLGRCSV RVSVNTANVT LGPQLMEVTV YRRHGRAYVP IAQVKDVYVV TDQIPVFVTM FQKNDRNSSD ETFLKDLPIM FDVLIHDPSH FLNYSTINYK WSFGDNTGLF VSTNHTVNHT YVLNGTFSLN LTVKAAAPGP CPPPPPPPRP SKPTPSLATT LKSYDSNTPG PAGDNPLELS RIPDENCQIN RYGHFQATIT IVEGILEVNI IQMTDVLMPV PWPESSLIDF VVTCQGSIPT EVCTIISDPT CEITQNTVCS PVDVDEMCLL TVRRTFNGSG TYCVNLTLGD DTSLALTSTL ISVP.

The protein encoded by this gene is a type I transmembrane glycoprotein which shows homology to the pMEL17 precursor, a melanocyte-specific protein. GPNMB shows expression in the lowly metastatic human melanoma cell lines and xenografts but does not show expression in the highly metastatic cell lines. GPNMB may be involved in growth delay and reduction of metastatic potential. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPNMB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene GPNMB
Gene Accession No. Q14956, Q6P7C7, Q99P91
Gene Alias Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; Gpnmb; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; Hgfin; ipd; iris pigment dispersion; NMB; Osteoactivin; si:ch211-155k24.6; transmembrane glycoprotein HGFIN; transmembrane glycoprotein NMB; Unknown (protein for MGC:133733); UNQ1725/PRO9925
Gene Symbols GPNMB
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10457, 113955, 93695
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.