Learn More
GPNMB Rabbit anti-Human, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | GPNMB |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | GPNMB |
| Gene Accession No. | Q14956 |
| Gene Alias | BOS_4109; Dchil; DC-HIL; dendritic cell-associated heparin sulfate proteoglycan-dependent integrin ligand; Dendritic cell-associated transmembrane protein; glycoprotein (transmembrane) nmb; glycoprotein (transmembrane) nmb-like; glycoprotein nmb; glycoprotein nmb-like protein; glycoprotein nonmetastatic melanoma protein B; GPNMB; hematopoietic growth factor inducible neurokinin-1; Hematopoietic growth factor inducible neurokinin-1 type; Hgfin; ipd; iris pigment dispersion; LOC100350570; Nmb; Osteoactivin; si:ch211-155k24.6; transmembrane glycoprotein HGFIN; Transmembrane glycoprotein NMB; UNQ1725/PRO9925 |
| Gene Symbols | GPNMB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.