Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533598
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533598 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533598 Supplier Invitrogen™ Supplier No. PA533598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%) Monkey, Marmoset (95%) Hamster, Dog (84%). This antibody is not recommended for use in ICC/IF.

The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1,000 members and is the largest known class of molecular targets with proven therapeutic value. GPR1 is an orphan receptor for the inflammation-associated leukocyte chemoattractant chemerin, suggesting a role for this receptor in the regulation of inflammation. In yeast, G protein-coupled receptor 1 senses glucose and controls filamentous growth. It acts upstream of adenylate cyclase and is required for glucose activation of cAMP synthesis in concert with a glucose phosphorylation-dependent mechanism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GPR1
Gene Accession No. P46091
Gene Alias Chemerin receptor 2; CMKLR2; G protein-coupled receptor 1; GPR1; Gpr-1; G-protein coupled receptor 1; probable G-protein coupled receptor 1
Gene Symbols GPR1
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GPR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2825
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.