Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102242 Supplier Invitrogen™ Supplier No. PA5102242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PRLHR.

PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH) that is highly expressed in anterior pituitary (Ozawa et al., 2002).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRLHR
Gene Accession No. P49683, Q64121, Q6VMN6
Gene Alias G protein-coupled receptor 10; Gm339; GPR10; G-protein coupled receptor 10; GR3; hGR3; MGC126539; MGC126541; Prlhr; prolactin releasing hormone receptor; prolactin releasing peptide receptor; prolactin-releasing hormone receptor; prolactin-releasing peptide receptor; PrRP receptor; Prrpr; UHR-1
Gene Symbols PRLHR
Host Species Rabbit
Immunogen A synthesized peptide derived from human PRLHR(Accession P49683), corresponding to amino acid residues L192-L242.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226278, 246075, 2834
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.