Learn More
Invitrogen™ GPR10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532993
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Rat (100%); Monkey, Marmoset, Mouse, Dog, Panda (95%); Bovine, Pig (89%); Horse (84%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
PRLHR is a 7-transmembrane domain receptor for prolactin-releasing hormone (PRLH) that is highly expressed in anterior pituitary (Ozawa et al., 2002).
Specifications
GPR10 | |
Polyclonal | |
Unconjugated | |
PRLHR | |
G protein-coupled receptor 10; Gm339; GPR10; G-protein coupled receptor 10; GR3; hGR3; MGC126539; MGC126541; Prlhr; prolactin releasing hormone receptor; prolactin releasing peptide receptor; prolactin-releasing hormone receptor; prolactin-releasing peptide receptor; PrRP receptor; Prrpr; UHR-1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
246075, 2834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P49683, Q64121 | |
PRLHR | |
Synthetic 19 amino acid peptide from 2nd extracellular domain of human PRLHR / GPR10. | |
50 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.