Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR101 Antibody (909603), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB7495PECY7
Description
GPR101 Monoclonal antibody specifically detects GPR101 in Human samples. It is validated for Flow CytometrySpecifications
GPR101 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
909603 | |
Flow Cytometry | |
G protein-coupled receptor 101, GPCR6, probable G-protein coupled receptor 101 | |
NS0 mouse myeloma cell line transfected with human GPR101, Met1-Pro508,, Accession # NP_473362 | |
0.1 mL | |
GPCR | |
83550 | |
Store at 4C in the dark. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction