Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR103 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31737625UL
Description
GPR103 Polyclonal antibody specifically detects GPR103 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GPR103 | |
Polyclonal | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
AQ27, G protein-coupled receptor 103, GPR103, G-protein coupled receptor 103, MGC149217, Orexigenic neuropeptide QRFP receptor, pyroglutamylated RFamide peptide receptor, QRFP receptor, SP9155 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: CIVNKTFSPAQRHGNSGITMMRKKAKFSLRENPVEETKGEAFSDGNIEVKLCEQTEEKKKLKRHLALFRSELAEN | |
25 μg | |
GPCR | |
84109 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction