Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR105 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981883
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1883 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1883 Supplier Bioss Supplier No. BS12028R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR105 has been reported to be expressed in brain. ESTs have been isolated from kidney, placenta, uterus, and heart/melanocyte/uterus libraries. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This 'turn-off' activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.

Specifications

Antigen GPR105
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene P2RY14
Gene Accession No. Q15391
Gene Alias A330108O13Rik; BPR105; G protein coupled receptor for UDP-glucose; G protein-coupled receptor 105; G protein-coupled receptor VTR 15-20; GPCR105; Gpr105; G-protein coupled receptor 105; KIAA0001; P2RY14; P2Y purinoceptor 14; P2Y(14) receptor; P2Y14; P2Y14 receptor; purinergic receptor P2Y, G-protein coupled, 14; purinergic receptor P2Y14; UDP-glucose receptor; VTR 15-20; VTR 15-20 receptor
Gene Symbols P2RY14
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human G-protein coupled receptor 105.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9934
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.