Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR107 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556822
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556822 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556822 Supplier Invitrogen™ Supplier No. PA556822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFRPASDNPY LQLSQEEEDL EMESVVTTSG VMESMKKVKK VTNGSVEPQG EWEGSV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.

GPR107 also named as KIAA1624 or LUSTR1 is 600 amino acid protein, which belongs to the LU7TM family. GPR107 localizes in membrane and is a multi-pass membrane protein. GPR107 is a promising candidate receptor for neuronostatin, and neuronostatin, interacting with GPR107, may play an important role in the central control of cardiovascular function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR107
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gpr107
Gene Accession No. Q5VW38
Gene Alias 9930033D15Rik; AI790205; bA138E2.2; C530034M11; G protein-coupled receptor 107; GCDRP; Gpr107; KIAA1624; Lung seven transmembrane receptor 1; LUSTR1; mKIAA1624; Protein GPR107
Gene Symbols Gpr107
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR107. Recombinant protein control fragment (Product #RP-95651).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.