Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR107 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9806 Supplier Bioss Supplier No. BS16284R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR107 also named as KIAA1624 or LUSTR1 is 600 amino acid protein, which belongs to the LU7TM family. GPR107 localizes in membrane and is a multi-pass membrane protein. GPR107 is a promising candidate receptor for neuronostatin, and neuronostatin, interacting with GPR107, may play an important role in the central control of cardiovascular function.

Specifications

Antigen GPR107
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene Gpr107
Gene Accession No. Q5VW38
Gene Alias 9930033D15Rik; AI790205; bA138E2.2; C530034M11; G protein-coupled receptor 107; GCDRP; Gpr107; KIAA1624; Lung seven transmembrane receptor 1; LUSTR1; mKIAA1624; Protein GPR107
Gene Symbols Gpr107
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR107.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57720
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.