Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR108 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102080
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102080 Supplier Invitrogen™ Supplier No. PA5102080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR108.

GPR108 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR108
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GPR108
Gene Accession No. Q9NPR9
Gene Alias 1810015L19Rik; AA589464; C79132; G protein-coupled receptor 108; GPR108; Lung seven transmembrane receptor 2; LUSTR2; Protein GPR108
Gene Symbols GPR108
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPR108(Accession Q9NPR9), corresponding to amino acid residues F496-L543.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56927
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.