Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR110 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100954
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100954 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100954 Supplier Invitrogen™ Supplier No. PA5100954
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR110.

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G protein coupled receptors translate extracellular signals into intracellular signals (G protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. GPR110 (G protein-coupled receptor 110), also known as PGR19, is a 911 amino acid protein that belongs to the G-protein coupled receptor 2 family and LN-TM7 subfamily. Characterized as an adhesion GPCR, GPR110 is a multipass membrane-bound protein with a long amino-terminus that contains multiple domains. One of these domains is the GPCR proteolytic site (GPS), which is essential for proteolytic cleavage of the amino-terminus and for cell surface expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR110
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRF1
Gene Accession No. Q5T601, Q8VEC3
Gene Alias 5031409J19Rik; Adgrf1; adhesion G protein-coupled receptor F1; adhesion G-protein coupled receptor F1; G protein-coupled receptor 110; G protein-coupled receptor KPG_012; G protein-coupled receptor PGR19; Gpr110; G-protein coupled receptor 110; G-protein coupled receptor KPG_012; hGPCR36; internal gene name of KIRIN laboratory:M5; KPG_009; KPG_012; PGR19; probable G-protein coupled receptor 110; seven transmembrane helix receptor
Gene Symbols ADGRF1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRF1(Accession Q5T601), corresponding to amino acid residues G838-H888.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266977, 77596
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.