Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR111 Antibody (594522), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB64941MFV610
Description
GPR111 Monoclonal antibody specifically detects GPR111 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
GPR111 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
0.1 mL | |
GPCR | |
222611 | |
Store at 4C in the dark. |
Western Blot, Flow Cytometry, CyTOF | |
594522 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
G protein-coupled receptor 111 | |
Chinese Hamster Ovary cell line, CHO-derived human GPR111, Cys19-Lys375, Accession # Q8IZF7 | |
RUO | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction