Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR111 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547845
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547845 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547845 Supplier Invitrogen™ Supplier No. PA547845
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) GPR30, rhGPR56, rhGPR114, rhGPR115, rhGPR124, and rhGPR125 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR111
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene ADGRF2
Gene Accession No. Q8IZF7
Gene Alias ADGRF2; adhesion G protein-coupled receptor F2; adhesion G-protein coupled receptor F2; G protein-coupled receptor 111; GPR111; G-protein coupled receptor 111; G-protein coupled receptor PGR20; hGPCR35; PGR20; probable G-protein coupled receptor 111
Gene Symbols ADGRF2
Host Species Sheep
Immunogen CHO-derived recombinant human GPR111 Cys19-Ser375.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222611
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.