Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR111 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116180
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116180 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116180 Supplier Invitrogen™ Supplier No. PA5116180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR111 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR111
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ADGRF2
Gene Accession No. Q8IZF7
Gene Alias ADGRF2; adhesion G protein-coupled receptor F2; adhesion G-protein coupled receptor F2; G protein-coupled receptor 111; GPR111; G-protein coupled receptor 111; G-protein coupled receptor PGR20; hGPCR35; PGR20; probable G-protein coupled receptor 111
Gene Symbols ADGRF2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 366-393 amino acids from the Central region of human GP111.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222611
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.