Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR112 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557228
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557228 Supplier Invitrogen™ Supplier No. PA557228
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: INTGKSQWEK PKFKQCKLLQ ELPDKIVDLA NITISDENAE DVAEHILNLI NESPALGKEE TKIIVSKISD ISQCDEISMN LTHVMLQIIN VVLEKQ Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 27%.

G protein-coupled receptors(GPRs or GPCRs), also known asseven transmembrane receptors, heptahelical receptors or 7TMreceptors, aremembers of the largest protein family and play a role in many differentstimulus-response pathways. G protein-coupled receptors mediate extracellularsignals into intracellularsignals(G protein activation). Theyrespond to awide variety of signaling molecules, including hormones, neurotransmitters and other proteins and peptides. GPR proteins are usuallyintegralseven passmembrane proteins with some conserved amino acid regions. GPR112 (G protein-coupled receptor 112) is a 3,080 amino acid multi-pass membrane protein that belongsto the G-protein coupled receptor 2 family and LN-TM7 subfamily. Localizing to cell membrane, GPR112 is highly expressed in normal enterochromaffin cells, aswell as neuroendocrine and primarylivercarcinoma. GPR112 contains one GPS domain andmayfunction as an orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR112
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRG4
Gene Accession No. Q8IZF6
Gene Alias ADGRG4; adhesion G protein-coupled receptor G4; adhesion G-protein coupled receptor G4; G protein-coupled receptor 112; Gm367; GPR112; G-protein coupled receptor 112; PGR17; probable G-protein coupled receptor 112; RP1-299I16
Gene Symbols ADGRG4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR112. Recombinant protein control fragment (Product #RP-94997).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.