Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR113 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560730 Supplier Invitrogen™ Supplier No. PA560730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CIPSTNLAYT AAWSPGEGSK ASSFNESGSQ CFVLAVQRCP MADTTYACDL QSLGLAPLRV PISITIIQDG DITCPEDASV LTWNVTKAGH VAQAPCPESK RGIVRRLCGA DG Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 70%.

Orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR113
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADGRF3
Gene Accession No. Q8IZF5
Gene Alias ADGRF3; adhesion G protein-coupled receptor F3; adhesion G-protein coupled receptor F3; G protein-coupled receptor 113; G protein-coupled receptor PGR23; Gm1041; GPR113; G-protein coupled receptor 113; G-protein coupled receptor PGR23; Pcr113; PGR23; probable G-protein coupled receptor 113; seven transmembrane helix receptor; UNQ9196/PRO34000
Gene Symbols ADGRF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GPR113. Recombinant protein control fragment (Product #RP-89581).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165082
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.