Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GPR113 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102081
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102081 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102081 Supplier Invitrogen™ Supplier No. PA5102081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GPR113.

Orphan receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR113
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ADGRF3
Gene Accession No. Q8IZF5
Gene Alias ADGRF3; adhesion G protein-coupled receptor F3; adhesion G-protein coupled receptor F3; G protein-coupled receptor 113; G protein-coupled receptor PGR23; Gm1041; GPR113; G-protein coupled receptor 113; G-protein coupled receptor PGR23; Pcr113; PGR23; probable G-protein coupled receptor 113; seven transmembrane helix receptor; UNQ9196/PRO34000
Gene Symbols ADGRF3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADGRF3(Accession Q8IZF5), corresponding to amino acid residues S30-G80.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165082
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.