Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR113 Rabbit anti-Human, Non-human Primate, Polyclonal, FabGennix

Catalog No. 502207267
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7267 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7267 Supplier Fabgennix Inc Supplier No. GPR113101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GPR113 Polyclonal antibody specifically detects GPR113 in Human, Non-human Primate samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen GPR113
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene ADGRF3
Gene Accession No. Q8IZF5
Gene Alias ADGRF3; adhesion G protein-coupled receptor F3; adhesion G-protein coupled receptor F3; G protein-coupled receptor 113; G protein-coupled receptor PGR23; Gm1041; GPR113; G-protein coupled receptor 113; G-protein coupled receptor PGR23; Pcr113; PGR23; probable G-protein coupled receptor 113; seven transmembrane helix receptor; UNQ9196/PRO34000
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 100-150 on human GPR113 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165082, 739846
Target Species Human, Non-human Primate
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.