Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR113 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983714
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3714 Supplier Bioss Supplier No. BS15375R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Orphan receptor.

Specifications

Antigen GPR113
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ADGRF3
Gene Accession No. Q8IZF5
Gene Alias ADGRF3; adhesion G protein-coupled receptor F3; Adhesion G-protein coupled receptor F3; G protein-coupled receptor 113; G protein-coupled receptor PGR23; Gm1041; GPR113; G-protein coupled receptor 113; G-protein coupled receptor PGR23; Pcr113; PGR23; probable G-protein coupled receptor 113; seven transmembrane helix receptor; UNQ9196/PRO34000
Gene Symbols ADGRF3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GPR113.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165082
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.